Learn More
Copper(II) sulfate pentahydrate, ACS, 98.0-102.0%
Description
Copper(II) sulfate, pentahydrate can be added to potassium permanganate to give an oxidant for the conversion of primary alcohols. It is used to etch zinc or copper plates for intaglio printmaking and as a mordant in vegetable dyeing. In analytical chemistry, it is used in Fehling's solution and Benedict's solution to test for reducing sugars. Also, it is used to test protein in Biuret test.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Crystalline |
| Quantity | 500 g |
| Assay Percent Range | 98.0-102.0% |
| Solubility Information | Soluble in water,methanol and glycerol. |
| Formula Weight | 249.68 (159.60 Anhydrous) |
| Grade | ACS |
| Density | 2.27 |
| Chemical Name or Material | Copper(II) sulfate, pentahydrate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
| Lot Number | Certificate Type | Date | Product Code |
|---|---|---|---|
| S20L022 | Certificate of Analysis | 22/06/2025 | 014178.A3,014178.36 |
| Q21L003 | Certificate of Analysis | 17/05/2025 | 014178.A3,014178.36 |
| P11L059 | Certificate of Analysis | 14/03/2025 | 014178.A3,014178.36 |
| N02C036 | Certificate of Analysis | 26/11/2024 | 014178.A3,014178.36 |
| W16K021 | Certificate of Analysis | 22/10/2024 | 014178.A3,014178.36 |
Safety and Handling
- Copper(II) sulfate pentahydrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Copper(II) sulfate pentahydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
