missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) sulfate pentahydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., 99-102%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
50 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 110°C (decomposition)(literature) |
CAS Max % | 100.0000% |
Quantity | 50 kg |
Assay Percent Range | 99 to 102% |
Linear Formula | KHSO4 |
UN Number | UN3077 |
Formula Weight | 249.69 |
Percent Purity | 99 to 102% |
Grade | ACS Reagent/Reagent ISO/Reagent Ph. Eur. |
Packaging | Fiberboard Box |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'