missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) tetrafluoroborate hexahydrate, 98%, Thermo Scientific Chemicals

Product Code. 11382658
Click to view available options
Quantity:
100 g
500 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72259-10-0
Molecular Formula B2CuF8H12O6
Molecular Weight (g/mol) 345.24
MDL Number MFCD05863735
InChI Key FBYIXFBIARQMJJ-UHFFFAOYSA-N
Synonym copper ii tetrafluoroborate hexahydrate, copper 2+ hexahydrate ditetrafluoroborate, copper ditetrafluoroborate hexahydrate, copper 2+ tetrafluoroborate-water 1/2/6, copper 2+ ion hexahydrate ditetrafluoroborate, borate 1-,tetrafluoro-, copper 2+ 2:1 , hexahydrate 9ci
PubChem CID 15774553
IUPAC Name copper(2+) bis(tetrafluoroboranuide) hexahydrate
SMILES O.O.O.O.O.O.[Cu++].F[B-](F)(F)F.F[B-](F)(F)F

Specifications

Color Blue
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 998%
Solubility Information Soluble in water.
Formula Weight 345.25 (237.15 Anhydrous)
Sensitivity Hygroscopic
Appearance Blue crystals
Chemical Name or Material Copper(II) tetrafluoroborate hexahydrate, hexahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'