Learn More
Copper(II) tetrafluoroborate hexahydrate, 98%, Thermo Scientific Chemicals
£59.90 - £235.00
Chemical Identifiers
CAS | 72259-10-0 |
---|---|
Molecular Formula | B2CuF8H12O6 |
Molecular Weight (g/mol) | 345.24 |
MDL Number | MFCD05863735 |
InChI Key | FBYIXFBIARQMJJ-UHFFFAOYSA-N |
Synonym | copper ii tetrafluoroborate hexahydrate, copper 2+ hexahydrate ditetrafluoroborate, copper ditetrafluoroborate hexahydrate, copper 2+ tetrafluoroborate-water 1/2/6, copper 2+ ion hexahydrate ditetrafluoroborate, borate 1-,tetrafluoro-, copper 2+ 2:1 , hexahydrate 9ci |
PubChem CID | 15774553 |
IUPAC Name | copper(2+) bis(tetrafluoroboranuide) hexahydrate |
SMILES | O.O.O.O.O.O.[Cu++].F[B-](F)(F)F.F[B-](F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11382658
|
Thermo Scientific Alfa Aesar
026127.22 |
100 g |
£59.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11372658
|
Thermo Scientific Alfa Aesar
026127.36 |
500 g |
£235.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
72259-10-0 | |
345.24 | |
FBYIXFBIARQMJJ-UHFFFAOYSA-N | |
15774553 | |
O.O.O.O.O.O.[Cu++].F[B-](F)(F)F.F[B-](F)(F)F |
B2CuF8H12O6 | |
MFCD05863735 | |
copper ii tetrafluoroborate hexahydrate, copper 2+ hexahydrate ditetrafluoroborate, copper ditetrafluoroborate hexahydrate, copper 2+ tetrafluoroborate-water 1/2/6, copper 2+ ion hexahydrate ditetrafluoroborate, borate 1-,tetrafluoro-, copper 2+ 2:1 , hexahydrate 9ci | |
copper(2+) bis(tetrafluoroboranuide) hexahydrate |
Specifications
72259-10-0 | |
Crystalline | |
B2CuF8H12O6 | |
copper ii tetrafluoroborate hexahydrate, copper 2+ hexahydrate ditetrafluoroborate, copper ditetrafluoroborate hexahydrate, copper 2+ tetrafluoroborate-water 1/2/6, copper 2+ ion hexahydrate ditetrafluoroborate, borate 1-,tetrafluoro-, copper 2+ 2:1 , hexahydrate 9ci | |
FBYIXFBIARQMJJ-UHFFFAOYSA-N | |
copper(2+) bis(tetrafluoroboranuide) hexahydrate | |
15774553 | |
Hygroscopic | |
Copper(II) tetrafluoroborate hexahydrate, hexahydrate |
Blue | |
998% | |
MFCD05863735 | |
Soluble in water. | |
O.O.O.O.O.O.[Cu++].F[B-](F)(F)F.F[B-](F)(F)F | |
345.24 | |
345.25 (237.15 Anhydrous) | |
Blue crystals |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: III
missing translation for 'einecsNumber' : 253-959-4
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only