missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Nickel wire, 0.075mm (0.003in) dia, Thermo Scientific Chemicals
Resistance alloy, negative lead for type J thermocouple
Specifications
Chemical Name or Material | Copper Nickel wire |
---|---|
Name Note | 0.075mm (0.003 in.) dia. |
Avg. Mol. Wt. or Mol. Wt. Range | Cu:Ni 55:45 wt% |
MDL Number | MFCD00801098 |
Synonym | Constantan |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11339378
|
Thermo Scientific Alfa Aesar
043269.H4 |
100 m |
N/A
|
N/A | |||||
11329378
|
Thermo Scientific Alfa Aesar
043269.H9 |
500 m |
N/A
|
N/A | |||||
Description
Resistance alloy, negative lead for type J thermocouple
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Copper Nickel wire | |
Cu:Ni 55:45 wt% | |
Constantan | |
Wire |
0.075mm (0.003 in.) dia. | |
MFCD00801098 | |
≈0.04g/m |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only