missing translation for 'onlineSavingsMsg'
Learn More
Copper powder, -100 mesh, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11344637

Copper powder, -100 mesh, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11344637
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11344637

Brand: Thermo Scientific Alfa Aesar 039682.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Color Red Brown
UN Number UN3089
Quantity 25 g
Percent Purity 99.999%
Mesh Size −100 Mesh
Odor Odorless
Form Powder
Assay (metals basis)
Chemical Name or Material Copper powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW26K036 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog Number039682.14,039682.22,039682.06
Lot NumberQ11K013 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number039682.14,039682.22,039682.06
Lot NumberX21I022 Certificate TypeCertificate of Analysis Date27/10/2022 Catalog Number039682.14,039682.22,039682.06,039690.18
Lot NumberM05I016 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number039682.14,039682.22,039682.06,039690.18
Lot NumberQ13G032 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number039682.14,039682.22,039682.06,039690.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper powder, -100 mesh, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.