missing translation for 'onlineSavingsMsg'
Learn More

Copper powder, -325 mesh, 10% max +325 mesh, 99% (metals basis)

Product Code. 11331509
Click to view available options
Quantity:
50 g
500 g
2 kg
10 kg
Unit Size:
10kg
2kg
500g
50g
This item is not returnable. View return policy

Product Code. 11331509

Brand: Thermo Scientific Alfa Aesar 013990.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Quantity 50 g
Mesh Size −325 Mesh 10% max +325 Mesh
Odor Odorless
Particle Size APS 8 to 11 Micron (FSSS)
Form Powder
UN Number UN3089
Percent Purity 99%
Assay (metals basis)
Chemical Name or Material Copper powder

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.