Learn More
Copper powder, -625 mesh, APS 2-3.50 micron, 99% (metals basis), Thermo Scientific Chemicals
£55.28 - £339.00
Chemical Identifiers
CAS | 7440-50-8 |
---|---|
Molecular Formula | Cu |
Molecular Weight (g/mol) | 63.55 |
MDL Number | MFCD00010965 |
InChI Key | RYGMFSIKBFXOCR-UHFFFAOYSA-N |
Synonym | cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer |
PubChem CID | 23978 |
ChEBI | CHEBI:30052 |
IUPAC Name | copper |
SMILES | [Cu] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11366597
|
Thermo Scientific Alfa Aesar
000622.09 |
10 g |
£55.28
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376597
|
Thermo Scientific Alfa Aesar
000622.18 |
50 g |
£134.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11386597
|
Thermo Scientific Alfa Aesar
000622.30 |
250 g |
£339.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-50-8 | |
63.55 | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
23978 | |
copper |
Cu | |
MFCD00010965 | |
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
CHEBI:30052 | |
[Cu] |
Specifications
7440-50-8 | |
8.94g/cm3 | |
Cu | |
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
[Cu] | |
63.55 | |
CHEBI:30052 | |
Odorless | |
Powder |
Copper colored powder | |
99% (Metals basis) | |
MFCD00010965 | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
copper | |
23978 | |
-625 Mesh | |
0 hPa | |
Copper |
Safety and Handling
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335-H500
missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: III
missing translation for 'einecsNumber' : 231-159-6
missing translation for 'rtecsNumber' : GL5325000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Copper powder
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H228-Flammable solid.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only