missing translation for 'onlineSavingsMsg'
Learn More

Copper, powder, 99+%, Thermo Scientific Chemicals

£19.50 - £162.81

Specifications

CAS 7440-50-8
Melting Point 1083°C
Boiling Point 2595°C
Molecular Formula Cu
Percent Purity ≥99%
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11309236
View Documents
Thermo Scientific Alfa Aesar
A16243.22
100 g
£19.50
100g
Please to purchase this item. Need a web account? Register with us today!
15453127
View Documents
Thermo Scientific Alfa Aesar
A16243.36
500 g
£52.56
500g
Please to purchase this item. Need a web account? Register with us today!
11319236
View Documents
Thermo Scientific Alfa Aesar
A16243.0E
2500 g
£162.81
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

7440-50-8
2595°C
≥99%
1083°C
Cu
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper, powder, 99+%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.