missing translation for 'onlineSavingsMsg'
Learn More

Copper silicide, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11357577
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Copper silicide thin film is used for passivation of copper-based chips, where it serves to suppress diffusion and electromigration and serves as a diffusion barrier. It is involved in the direct process, the industrial route to organosilicon compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12159-07-8
Molecular Formula Cu5Si
MDL Number MFCD00134719

Specifications

Melting Point 825°C
Color Silver
Physical Form Pieces
Quantity 25 g
Assay Percent Range 99.5% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 345.79
Odor Odorless
Chemical Name or Material Copper silicide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'