missing translation for 'onlineSavingsMsg'
Learn More

Copper Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™

£63.05

Chemical Identifiers

CAS 3251-23-8
Molecular Formula CuN2O6
Molecular Weight (g/mol) 187.554
MDL Number 10967
InChI Key XTVVROIMIGLXTD-UHFFFAOYSA-N
Synonym copper dinitrate, cupric nitrate, copper ii nitrate, copper nitrate, cupric dinitrate, claycop, copper 2+ nitrate, caswell no. 246, nitric acid, copper 2+ salt, unii-9tc879s2zv
PubChem CID 18616
ChEBI CHEBI:78036
IUPAC Name copper;dinitrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Cu+2]
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10418220
View Documents
Fisher Chemical
J/8025/08
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Copper Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M

Specifications

Chemical Identifiers

3251-23-8
187.554
XTVVROIMIGLXTD-UHFFFAOYSA-N
18616
copper;dinitrate
CuN2O6
10967
copper dinitrate, cupric nitrate, copper ii nitrate, copper nitrate, cupric dinitrate, claycop, copper 2+ nitrate, caswell no. 246, nitric acid, copper 2+ salt, unii-9tc879s2zv
CHEBI:78036
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Cu+2]

Specifications

Colorless
3251-23-8
10967
XTVVROIMIGLXTD-UHFFFAOYSA-N
copper;dinitrate
18616
HDPE plastic bottle
1
CuN2O6
copper dinitrate, cupric nitrate, copper ii nitrate, copper nitrate, cupric dinitrate, claycop, copper 2+ nitrate, caswell no. 246, nitric acid, copper 2+ salt, unii-9tc879s2zv
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Cu+2]
187.554
CHEBI:78036
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Copper solution 1000 ppm

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS
Documents

Documents

Product Certifications