missing translation for 'onlineSavingsMsg'
Learn More

Copper Standard Solution, 0.1%, Reagecon™
BIOPHARMA

Product Code. 12812503
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
Packaging:
100mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Concentration 0.001
Packaging 100mL
Quantity 100 mL
Product Identifier
  • COPPER STANDARD SOLUTION (0.1% Cu)
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : copper sulphate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'