missing translation for 'onlineSavingsMsg'
Learn More

Copper for AAS 1g/L HNO3 matrix, Fisher Chemical™

Product Code. 12983265
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Concentration 1g/L ±0.002g/L
Packaging Bottle
Quantity 1000 mL
Type Flame Single-element Standard Solution
UN Number 3082
Solvent or Matrix 2% HNO3
Physical Form Liquid
Chemical Name or Material Copper
compliance-icons
Product Identifier
  • Single-element Standard Solution for AAS. Copper (Cu) 1000mg/l in HNO3 2%
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'