missing translation for 'onlineSavingsMsg'
Learn More

Copper wire, 0.5mm (0.02in) dia, annealed, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11366877
Click to view available options
Quantity:
100 m
250 m
500 m
This item is not returnable. View return policy
This item is not returnable. View return policy

Form: approx. 1.75g/m; Temper: soft

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Appearance Copper colored wire
Density 8.94g/cm3
Assay Percent Range 99.9% (Metals basis)
Quantity 100 m
Formula Weight 63.546
Odor Odorless
Vapor Pressure 0 hPa
Form Wire
Chemical Name or Material Copper, Annealed

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'