missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cordycepin, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Nucleoside analog that acts as an anticancer and antifungal agent. Can be converted to 3'-deoxyadenosine triphosphate (3'-dATP), which inhibits ATP-dependent DNA synthesis. Inhibits growth of various tumor cells in vitro.
Specifications
CAS | 73-03-0 |
Molecular Formula | C10H13N5O3 |
MDL Number | MFCD00037998 |
Quantity | 10 mg |
Synonym | cordycepin, 3'-deoxyadenosine, cordycepine, 9-cordyceposidoadenine, adenosine, 3'-deoxy, 9-beta-d-3'-deoxyribofuranosyl adenine, unii-gz8vf4m2j8, 9h-purine, 6-amino-9-3-deoxy-beta-d-ribofuranosyl, ccris 3233, gz8vf4m2j8 |
InChI Key | OFEZSBMBBKLLBJ-PNFQIUPPNA-N |
SMILES | NC1=C2N=CN([C@@H]3O[C@H](CO)C[C@H]3O)C2=NC=N1 |
IUPAC Name | (2R,3R,5S)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolan-3-ol |
Molecular Weight (g/mol) | 251.25 |
PubChem CID | 6303 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'