Learn More
(-)-Corey lactone diol, MedChemExpress
MedChemExpress (-)-Corey lactone diol is a reduced version of corey aldehyde. A building block in the chemical synthesis.
Brand:  Medchem Express HY-W008393
Encompass_Preferred
Product Code. 16449336
  • £56.87 / 100mg
Explore more special offers
Description
Description
(-)-Corey lactone diol is a reduced version of corey aldehyde. A building block in the chemical synthesis.
Specifications
Specifications
(-)-Corey lactone diol
White to Off-White
C8H12O4
DMSO : 100 mg/mL (580.79 mM; Need ultrasonic)
172.18
97.0%
Solid
32233-40-2
Research
100 mg
OC[C@@H]([C@H](O)C[C@]1([H])O2)[C@@]1([H])CC2=O
172.18
Research