Learn More
R&L Enterprises™ Cork Borer Sharpner
Keeps cork borers sharp and efficient to use. R&L Enterprises™ Cork Borer Sharpeners are for cork borers from 4 to 27mm.
Brand: R&L Enterprises™ 3403
Code : 03
Additional Details : Weight : 0.22560kg
Description
- With metal handle
- Metal cone with hinged cutter blade set in slot
Specifications
Cork Borer Sharpner | |
For 4 to 27mm dia. Borers | |
1Each |