Learn More
Chroma Gesellschaft™ Cork Borers
Efficiently and cleanly bores holes in cork. Chroma Gesellschaft™ Cork Borers have a cutting tube mounted with safety under-flange in shaped handle.
Brand: Chroma Gesellschaft™ 3400
Code : 03
Additional Details : Weight : 0.19260kg
Description
- With rod for clearing bores
- Manufactured from brass
- Sizes numbered consecutively from 1 upwards in each set
Specifications
Cork Borers | |
Brass | |
For boring holes in cork. | |
4 to 10 mm |