missing translation for 'onlineSavingsMsg'
Learn More

Corn oil

Product Code. 10208473 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10208473

Brand: Thermo Scientific Acros 405430025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 8001-30-7

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-30-7
MDL Number MFCD00130865

Specifications

Density 0.9200g/mL
Flash Point >200°C
Packaging Plastic Bottle
Refractive Index 1.4730 to 1.4760
Quantity 2.5 kg
Merck Index 14, 2533
Specific Gravity 0.92
Solubility Information Solubility in water: insoluble
Chemical Name or Material Corn oil

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.