missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 0.05% Trypsin/0.53mM EDTA in HBSS w/o Calcium, Magnesium or Sodium Bicarbonate
Used to enzymatically release adherent cells from tissue culture plates for passaging.
£114.55 - £222.83
Specifications
Content And Storage | -20°C |
---|---|
Form | Liquid |
Product Type | Cell Culture Dissociation Reagent |
Sterility | Sterile |
Without Additives | No Calcium, No Magnesium, No Sodium Bicarbonate |
Product Code | Brand | Quantity | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Product Type | Price | Quantity & Availability | ||||
15323651
|
Corning™
25-052-CV |
6 x 500 mL | Cell Culture Dissociation Reagent |
£222.83
Pack of 6 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15313651
|
Corning™
25-052-CI |
6 x 100 mL | Cell Culture Dissociation Reagent |
£114.55
Pack of 6 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Trypsin is used to enzymatically release adherent cells from tissue culture plates for passaging. Divalent cations such as calcium and magnesium, which are often present in the cell culture environment, inhibit this action. EDTA sequesters these ions and thus enhances the efficacy of trypsin. This product contains trypsin at a concentration of 0.05% with 0.53 mM EDTA, and does not contain sodium bicarbonate.Specifications
-20°C | |
Cell Culture Dissociation Reagent | |
No Calcium, No Magnesium, No Sodium Bicarbonate |
Liquid | |
Sterile | |
7 to 7.6 |