missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 0.25% Trypsin in HBSS w/o Calcium and Magnesium
Enzymatic dissociation reagents tested for porcine parvovirus
Brand: Corning™ 25-050-CI
Code : NEW
Additional Details : Weight : 0.60000kg
Specifications
-20°C | |
Cell Culture Dissociation Reagent | |
No Calcium, No Magnesium | |
6 x 100 mL |
Liquid | |
Sterile | |
7 to 7.6 |