missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 2.5% Trypsin in HBSS w/o Calcium, Magnesium and Phenol Red

Enzymatic dissociation reagents tested for porcine parvovirus

Brand:  Corning™ 25-054-CI

Code : NEW

Additional Details : Weight : 0.90000kg

Product Code. 15343651

  • £183.00 / Pack of 6
In Stock
to see stock.

Description

Description

Specifications

Specifications

-20°C
Cell Culture Dissociation Reagent
No Calcium, No Magnesium, No Phenol Red
6 x 100 mL
Liquid
Sterile
7 to 7.6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers