missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Untreated 245mm Square BioAssay Dishes
GREENER_CHOICEBIOPHARMA

Product Code. 10737041 Shop All Corning Products
Click to view available options
Dimensions (L x W x H):
245 x 245 x 20 mm
245 x 245 x 25 mm
245 x 245 x 26 mm
Unit Size:
Pack of 16
Pack of 20

Product Code. 10737041

Brand: Corning™ 431111

Only null left
Add to basket
Edge
Add to basket

Designed with a generous growth area of 500cm2 to allow for larger assays and better overall use of space in incubators. Corning™ Untreated 245mm Square Low Profile BioAssay Dishes provide a superior option for applications where cell attachment is not desired.

Improve the efficiency of microbial assays including library replication, colony picking, and plaque lift studies.

 

  • Polystyrene dishes feature stacking beads for stable stacking
  • Compatible with various automated colony picking and gridding robots
  • Supplied with vented lid to provide consistent gas exchange
  • Sterile, certified nonpyrogenic
  • Patented (U.S. Patent No.. 5,856,176) design allows for easier handling

 

TRUSTED_SUSTAINABILITY

Specifications

Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Bottom Shape Square
Capacity (Metric) 150 mL (Well)
Color Clear
Culture Area 500 sq. cm
Disposable Single-use
Material Polystyrene
Shape Square
Sterility Sterile
Type Cell Culture Dish
Includes Lid, Handles
Dimensions (L x W) 245 x 245 mm
Dimensions (L x W x H) 245 x 245 x 25 mm
Length (Metric) 245 mm
Width (Metric) 245 mm
Height (Metric) 25 mm
Depth (Metric) Inside 18 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.