Learn More
Corning™ 384-Well, Fibronectin-Treated, Flat-Bottom Microplate
Color:Clear; Sterility:Non-sterile; Lid:With Lid
Brand:  Corning™ 3847
Code : A0
Additional Details : Weight : 11.15836kg
Product Code. 13246768
  • £3041.04 / Pack of 100
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
Description
Description
  • Total well volume of 112 μL; working well volume of 20 μL – 80 μL
  • Cell culture microplates are sterile and nonpyrogenic.
  • The 384-well Universal Optics NBS microplate is manufactured using an advanced polymer with high clarity and improved chemical resistant properties.
  • Specifications
    Specifications
    Standard
    16 x 24
    384
    Fibronectin
    100/Cs.
    With Lid
    Clear
    Non-sterile
    Flat Bottom (F-well)
    20/Pk.