missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom Microplate
Color:White; Sterility:Sterile; Lid:With Lid
Brand: Corning™ 3570
Description
- Minimal well to well crosstalk
- White microplates enhance luminecent signals and have low background luminescence
- Black mircoplates have low background fluorescence and minimize light scattering
- Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
- Medical-grade polystyrene
- Well volume: 112μL
- Working volume: 20μL to 80μL
- Surface area/well: 0.06cm2
- Treated surface options are not coated, require no special handling and are stable at room temperature
- Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
- Plate dimensions meet proposed industry standards
Specifications
Standard | |
16 x 24 | |
White | |
384 | |
Sterile | |
Flat Bottom (F-well) | |
Lid |
With Lid | |
10/Pk., 50/Cs. | |
Polystyrene | |
20 to 80 μL | |
Cell Culture | |
Attachment and growth of anchorage-dependent cells | |
50/Cs. |