missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 384-Well Low-Volume Solid Microplates
Same low reagent volumes as 1536-well plates without the need for new instrumentation
£484.73 - £831.36
Specifications
Format | Standard |
---|---|
Array | 16 x 24 |
Material | Polystyrene |
No. of Wells | 384 |
Sterility | Non-sterile |
Product Code | Brand | Color | Surface Treatment | Volume (Metric) Working | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Surface Treatment | Volume (Metric) Working | Price | Quantity & Availability | |||
10679029
|
Corning™
3825 |
White | Non-treated | 5 to 40 μL |
£484.73
Pack of 50 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
10064094
|
Corning™
3824 |
White | Non-binding | 5 to 40 μL |
£509.69
Pack of 50 |
In Stock
Log in to see stock. |
|||
10583234
|
Corning™
3824BC |
White | Non-binding | 5 to 40 μL |
£788.25
Pack of 50 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
10699921
|
Corning™
3674 |
White | Non-treated | 5 to 40 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10167431
|
Corning™
3677 |
Black | Non-treated | 2 to 25 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10027781
|
Corning™
3676 |
Black | Tissue Culture | 2 to 20 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10342952
|
Corning™
3673 |
White | Non-binding | 5 to 40 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Polystyrene plates feature a unique square-to-round well design with a conical-shaped bottom that provides easier access and imaging for superior assay performance.
- Bottom reduces fluid and air entrapment, greater signal-to-noise ratio and reduced FP signal (mP) when working with very low volumes
- Each well has a raised rim to reduce cross-contamination during assay and liquid-handling process
- Choose black plates for fluorescence applications or white for luminescence applications
- Ideal for immunoassays
- Can bind medium (>10kDa) and large biomolecules that possess ionic groups and/or hydrophobic regions
- Working volume: 2 to 20μL
Specifications
Standard | |
Polystyrene | |
Non-sterile |
16 x 24 | |
384 |