Learn More
Corning™ 384-Well Solid Black or White Polystyrene Microplates

Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Solid Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
Brand: Corning™ 3820
465.94 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Designed to reduce well-to-well cross-talk during fluorescent and luminescent assays.

Specifications
Standard | |
(NBS) is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions. | |
Non Binding | |
Polystyrene | |
5 to 40 μL | |
Non-binding | |
Flat Bottom (F-well) | |
50/Cs. | |
50/Cs. |
Without Lid | |
16 x 24 | |
Black | |
384 | |
Non-sterile | |
50 μL | |
Reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise | |
10/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.