missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 384-Well Nonbinding Surface (NBS™) Microplates
Enhanced signal-to-noise ratio
£598.32 - £1300.09
Specifications
Format | Standard |
---|---|
Array | 16 x 24 |
For Use With (Equipment) | Plate readers, liquid handlers and robotic systems |
No. of Wells | 384 |
Volume (Metric) Working | 20 to 80 μL |
Product Code | Brand | Color | Well Color | Barcode | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Well Color | Barcode | Price | Quantity & Availability | |||
10266083
|
Corning™
3574BC |
White | White | Includes Barcode |
£604.47
Pack of 50 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||
10782684
|
Corning™
3575BC |
Black | Black | Includes Barcode |
£598.32
Pack of 50 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||
10317511
|
Corning™
3653 |
White, Clear Bottom | Clear | No Barcode |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10728402
|
Corning™
3655 |
Black | Clear | No Barcode |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11353725
|
Corning™
3653BC |
White | Clear | Includes Barcode |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Ideal for homogenous assays such as Scintillation Proximity Assays (SPA) and fluorescent assays
- Modified polymer surface is nonionic hydrophilic, minimizing bio-interaction within wells to reduce nonspecific binding and reagent costs
- Significantly reduces protein and nucleic acid binding to polymers, maintains enzyme activity, and inhibits adhesion of a number of cell lines Stable to aqueous solutions containing EtOH
- Use at temperature range of 4° to 37°C (39° to 99°F)
- Flat-bottom wells
- Compatible with a wide range of plate readers, liquid handlers, and robotic systems
Generic Bar Coded Plates
- Code 128 bar code format ensures compatibility with most bar code scanning and software systems
- To avoid mix-ups, the surface treatment of the microplate wells is easily identified
- Labels applied to all four sides of the plate No sequence number duplicates
Specifications
Standard | |
Plate readers, liquid handlers and robotic systems | |
20 to 80 μL | |
Non-binding | |
For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays |
16 x 24 | |
384 | |
Non-sterile | |
Flat Bottom (F-well) |