Learn More
Corning™ 384-Well, Ultra-Low Binding, U-Shaped-Bottom Microplate
GREENER_CHOICE
Color:Black, Clear Bottom; Sterility:Sterile
Brand:  Corning™ 4516
Product Code. 15354077
  • £282.00 / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
With their novel and proprietary design, these microplates are ideal for generating and analyzing 3D multicellular spheroids in the same microplate. The Ultra-Low Attachment surface enables uniform and reproducible 3D multicellular spheroid formation. The black opaque microplate body shields each optically clear, round bottom well from well-to-well cross-talk.
  • Optically clear round bottom with black opaque microplate body
  • Covalent attachment of Ultra-Low Attachment (ULA) surface to reduce cellular adhesion to well surface
  • Novel well geometry aids in the generation of uniform, single spheroids across all wells, which enables automated visualization.
  • Unique design shields each well to minimize well-to-well cross-talk.
  • You can culture and assay spheroids in the same microplate without the need for transfer to a new microplate.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
0.06 cm2
5/Pk.
Spheroid Microplate
384
Sterile
90 μL
Closure
1/Pk.
With Lid
16 x 24
Black, Clear Bottom
Polystyrene
20 to 80 μL
Ultra-low Binding
Round Bottom (U-well)
5/Cs.