Learn More
Corning™ 12-Strip, 96-Well, High Binding, Flat-Bottom Microplate
GREENER_CHOICE
Color:Clear; Sterility:Non-sterile; Lid:Without Lid
Brand:  Corning™ 42592
Product Code. 15212359
  • £575.00 / Pack of 100
Estimated Shipment: 22-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Sold 100 per case.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
8 x 12
Clear
Non-sterile
Flat Bottom (F-well)
Without Lid
High Binding, 4CAV
96
High Binding
100/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ 12-Strip, 96-Well, High Binding, Flat-Bottom Microplate > 1 x 8; High binding; Clear; Flat bottom; Without lid; Nonsterile; 4 CAV