missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96 Half Area Well Flat Clear Bottom Black Polystyrene TC-treated Microplates, Without Lid, Sterile, 25/Bag
Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
Brand: Corning™ 3887A
Code : NEW
Additional Details : Weight : 6.17000kg
Description
- Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm
- Opaque walls prevent well-to-well cross-talk
- Optically clear flat bottom permits direct microscopic viewing
Specifications
Half-Area | |
0.16 cm2 | |
Black, Clear Bottom | |
Polystyrene | |
96 | |
Sterile | |
Flat Bottom (F-well) | |
25/Pk., 100/Cs. |
Without Lid | |
8 x 12 | |
Standard automated handling and detection systems | |
50 to 100 μL | |
25 to 125 μL | |
Cell Culture | |
Homogenous and cell based assays | |
Clear |