missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well Half Area Black Flat Bottom Polystyrene NBS™ Microplate

Designed to reduce well-to-well crosstalk
Brand: Corning™ 3993
240.82 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
Half-Area | |
8 x 12 | |
Polystyrene | |
25 to 125 μL | |
Non-binding | |
Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise | |
5/Pk. |
Without Lid | |
Black | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
25/Cs. | |
25/Cs. |
COR001 Lifescience Guid
File size: 7 MB
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
10Hours
7Minutes
33Seconds
51
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ 96-Well Half Area Black Flat Bottom Polystyrene NBS™ Microplate > Black; Half Area; No; NBS; Flat
Spot an opportunity for improvement?Share a Content Correction