missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Half Area Flat Bottom Polystyrene NBS™ Microplates
GREENER_CHOICE

Product Code. p-3319738 Shop All Corning Products
Click to view available options
Color:
White
White, Clear Bottom
Unit Size:
Pack of 100
Pack of 25
This item is not returnable. View return policy

Product Code. 10522063

Brand: Corning™ 3642

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed to reduce well-to-well crosstalk during luminescent assays

  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Format Half-Area
    Lid Without Lid
    Array 8 x 12
    Color White
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 25 to 125 μL
    Sterility Non-sterile
    Surface Treatment Non-binding
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.