Learn More
Corning™ 96-Well Next Generation CrystalEX Protein Crystallization Microplates
Optimized for high-throughput sitting-drop protein crystallization
Brand:  Corning™ 3552
 View more versions of this product
Product Code. 10057412
  • £690.00 / Pack of 50
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Corning™ 96-well and 384-well crystallization microplates are optimized for high throughput protein crystal growth and screening
  • Designed for sitting drop applications
  • Meets ANSI/SBS standard footprint dimensions for microplates
  • Ideal for fully automated crystal screening
  • Multiple formats and versatility for custom options to maximize crystal formation, identification and analysis, and harvesting
  • - Choose from four unique protein well shapes
    - Options include one or three protein wells per reservoir well
  • Cyclic olefin copolymer (COC) offers strong chemical compatibility and good optical clarity
  • Reservoir numbers are embossed on each individual well for easy identification
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Alphanumeric
Without Lid
Clear
2 μL
Non-sterile
Round, Flat Bottom (F-well)
10/Pk.
Standard
8 x 12
PZero
96
Non-treated
50/Cs.
50/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products