Learn More
Corning™ 96-Well Round Bottom Polystyrene Ulta-Low Attachment Microplate
Ulta-Low attachment surface with covalently bonded hyrogel minimizes cell attachment, protein absorption, enzyme activation and cellular activation
Brand:  Corning™ 4591
470.46 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Product Code. 17750708
  • £586.00 / Pack of 24
Estimated Shipment: 01-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Standard
    8 x 12
    Polystyrene
    96
    Sterile
    24/Cs.
    With Lid
    Black
    360 μL
    75 to 200 μL
    Round
    1/Pk.
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Corning™ 96-Well Round Bottom Polystyrene Ulta-Low Attachment Microplate > Black; Standard; Yes; Ultra Low Attachment; Round