missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well White Flat Bottom Polystyrene NBS Microplate Without Lid
Nonionic hydrophilic surface
Brand: Corning™ 3990
Code : 66
Additional Details : Weight : 1.72365kg
Description
Specifications
Standard | |
8 x 12 | |
Polystyrene | |
75 to 200 μL | |
Non-binding | |
Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise | |
5/Pk. |
Without Lid | |
White | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
25/Cs. | |
25/Cs. |