Learn More
Axygen™ Gel Documentation System Accessory, White Light Conversion Screen
For use with Gel Documentation Systems (GD-1000 and GDBL-1000)
Brand: Axygen™ GD-WLCS
This item is not returnable.
View return policy
Description
White light; For imaging of visible stains and dyes; Rnase/Dnase freeSpecifications
Gel Documentation Systems (GD-1000 and GDBL-1000) | |
White Light Conversion Screen |