Learn More
Corning™ Bar Coded Storage Tubes: 1D/2D Bar Coded
For enhanced sample storage, identification and retrieval
Description
Feature a synchronized coding scheme that minimizes confusion and the risk of duplication, while enabling safe and secure sample identification.
- Maximum Identification — Synchronized 2D and linear bar code
- Maximum Information — 14 x 14 dot 2D bar code
- Bar Code Stability — Laser-etched, not an attached label
- Compound Compatibility — Polypropylene construction throughout creates inert vessel
- Temp. tolerance: -180 to +110°C (screw cap); -80 to +110°C (TPE cap)
- Length: 43.9mm
- 96/rack
Specifications
Polypropylene | |
Non-sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Corning™ Bar Coded Storage Tubes: 1D/2D Bar Coded