Learn More
Corning™ Basic Fibroblast Growth Factors (bFGF), Human Recombinant
Isolated from pituitary glands
£203.32 - £1640.00
Specifications
Content And Storage | -20°C |
---|---|
Product Type | Growth Factor |
Formulation | Lyophilized from phosphate buffered saline, carrier free, reconstitute in dH2O or serum-free medium |
For Use With (Application) | Useful for the maintenance and differentiation of sympathetic and sensory neurons, as well as neuronal cells in-vitro |
Purity or Quality Grade | >95% by SDS-PAGE |
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
11563530
|
Corning™
354060 |
Growth Factor | 10 μg |
£203.32
10µg |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11503620
|
Corning™
356060 |
Growth Factor | 5 x 10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11513620
|
Corning™
356061 |
Growth Factor | 10 x 10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.
- Vialed, highly purified growth factors with high biological activity
- Broad range of highly purified natural and recombinant lymphokines
- Enriched culture supplements to be reconstituted/added in the medium of your choice
- Propagate your cells under more defined serum-reduced or serum-free conditions
Growth factors are proteins that bind to receptors found on the surface of non-hematopoietic cells. Each family of growth factors affect specific cell types, e.g., nerve growth factors (NGF) affect nerve cell types, and epidermal growth factors (EGF) affect epithelial cell types.
Corning offers a range of growth factors that are derived from different species, both natural and recombinant. These growth factors have been used to promote long-term proliferation in cell culture and to induce cell differentiation in a variety of cell types under serum-free or reduced serum conditions.
Order Info
- FGF is supplied lyophilized, bFGF in 3.6mM Tris, PBS pH 7 buffer
- bFGF (human) is lyophilized
Specifications
-20°C | |
Lyophilized from phosphate buffered saline, carrier free, reconstitute in dH2O or serum-free medium | |
>95% by SDS-PAGE | |
E. coli |
Growth Factor | |
Useful for the maintenance and differentiation of sympathetic and sensory neurons, as well as neuronal cells in-vitro | |
Human |