Learn More
Corning™ BioCoat™ #1 German Glass Coverslips, Round
Coated with a variety of extracellular matrix substrates. Corning™ BioiCoat™ No. 1 German Glass Coverslips provide an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures.
£225.00 - £324.00
Specifications
Type | Slide Coverslip |
---|---|
Material | No. 1 German Glass |
For Use With (Application) | Fluorescence imaging of live cells, confocal microscopy, high resolution and inverted microscopy, phase contrast microscopy |
Product Code | Brand | Diameter (Metric) | Surface Treatment | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Surface Treatment | Price | Quantity & Availability | ||||
10220772
|
Corning™
354085 |
12 mm | Poly-L Lysine |
£302.04
Pack of 80 |
In Stock
Log in to see stock. |
||||
10727911
|
Corning™
354086 |
12 mm | Poly-D Lysine |
£303.00
Pack of 80 |
In Stock
Log in to see stock. |
||||
10543322
|
Corning™
354088 |
22 mm | Fibronectin |
£274.00
Pack of 60 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
||||
10468681
|
Corning™
354087 |
12 mm | Laminin/Poly-D Lysine |
£324.00
Pack of 80 |
In Stock
Log in to see stock. |
||||
10553322
|
Corning™
354089 |
22 mm | Collagen Type I |
£225.00
Pack of 60 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
||||
Description
Corning™ BioCoat™ German Glass coverslips with surface treatment (see individual Specification tables for coating). Corning BioCoat products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
Shelf life: ≥ 3 months from date of shipment
Specifications
Slide Coverslip | |
No. 1 German Glass | |
Fluorescence imaging of live cells, confocal microscopy, high resolution and inverted microscopy, phase contrast microscopy |