Learn More
Corning™ BioCoat™ Angiogenesis System for Endothelial Cell Tube Formation
Evaluate and quantitate endothelial cell invasion using real-time fluorescence and non-destructive sample preparation
Brand: Corning™ 354150
Includes: A Falcon™ 24-Multiwell Insert Plate and a non-tissue culture-treated 24-well receiver plate with lid
Description
- Highly reproducible assay system
- Ideal for screening anti-angiogenic agents
- Rapid image acquisition and data quantification
Specifications
3 μm | |
5 | |
5/Cs. | |
Black, Clear |
Microplate | |
5/Pk. | |
BioCoat | |
96 |
Product Content Correction
Product Title
Corning™ BioCoat™ Angiogenesis System for Endothelial Cell Tube Formation > 5/Cs
Spot an opportunity for improvement?Share a Content Correction