Learn More
Corning™ BioCoat™ Angiogenesis System for Endothelial Cell Tube Formation
Evaluate and quantitate endothelial cell invasion using real-time fluorescence and non-destructive sample preparation
Brand: Corning™ 354149
Code : 67
Additional Details : Weight : 0.19000kg
Includes: A Falcon™ 24-Multiwell Insert Plate and a non-tissue culture-treated 24-well receiver plate with lid
Description
- Highly reproducible assay system
- Ideal for screening anti-angiogenic agents
- Rapid image acquisition and data quantification
Specifications
3 μm | |
1 | |
1/Ea. | |
Black, Clear |
Microplate | |
1/Pk. | |
BioCoat | |
96 |