missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ Assay System
Automation-friendly design
£363.80 - £1578.77
Specifications
Color | Clear |
---|---|
Culture Area | 0.3 cm2 |
Sterility | Non-sterile |
Surface Treatment | Collagen |
Well Count | 24 |
Includes: 24-Multiwell™ Insert plate, specialized media to perform 24 assays, feeder tray, and lid.
Product Code | Brand | Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Quantity | Price | Quantity & Availability | ||||
10631932
|
Corning™
354802 |
Cell Culture Insert System | 5/Cs. |
£1578.77
Pack of 5 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
||||
11513590
|
Corning™
354801 |
Cell Culture Insert System | 1/Cs. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10596951
|
Corning™
354804 |
Multi-well Insert System | 5/Cs. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
Specifications
Clear | |
Non-sterile | |
24 | |
1 μm |
0.3 cm2 | |
Collagen | |
24 multiwell insert plate, specialized media to perform 24 assays, feeder tray and lid |