missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ Collagen I Glass Multiwell Culture Slide
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
£375.00 - £420.00
Specifications
Type | Culture Slide |
---|---|
For Use With (Application) | Adhesion, growth and differentiation of neuronal and transfected cell lines |
No. per Pack | 3/Pk. |
No. per Case | 12/Cs. |
Surface Coating | Collagen I |
Product Code | Brand | No. of Wells | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | Price | Quantity & Availability | |||||
10595431
|
Corning™
354630 |
8 |
£375.00
Pack of 12 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||||
10219841
|
Corning™
354557 |
4 |
£420.00
Pack of 12 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||||
Description
Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.
Applications include:
- Promotion of cell attachment and spreading
- Rapid expansion of cell populations
- Serum-free or reduced serum culture
- Cell adhesion assays
- Improving survival of primary cells in culture
Source
Quality Control
- Tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
- Tested and found negative for bacteria and fungi
- Collagen I purity >90% by SDS-PAGE
Storage
Spécification
Culture Slide | |
3/Pk. | |
Collagen I |
Adhesion, growth and differentiation of neuronal and transfected cell lines | |
12/Cs. |