missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Collagen I Inserts with PET Membrane

Product Code. 10366151
Click to view available options
Pore Size:
0.3 μm
0.4 μm
0.45 μm
1 μm
3 μm
Type:
Cell Culture Insert
Well Plate
Quantity:
1
2
4
Unit Size:
Pack of 24
Pack of 4
This item is not returnable. View return policy

Product Code. 10366151

Brand: Corning™ 354442

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines

  • Each lot is tested for the ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
  • Tested and found negative for bacteria and fungi
  • Cellware is also tested for ability to promote firm attachment of rat cerebellar granule cells
  • Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
  • Matrix components are applied as an even, optically clear coating
  • All lots tested and found negative for bacteria and fungi; cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells

For research use only.

Order Info

Bulk quantities are also available; contact your Fisher Scientific Customer Service Representative.

TRUSTED_SUSTAINABILITY

Specifications

Inserts per Plate 6
Material PET
Pore Size 0.45 μm
Type Well Plate
For Use With (Application) Adhesion, growth and differentiation of neuronal and transfected cell lines
Quantity 4
Storage Requirements 2° to 8°C
No. of Wells 6
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.