missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ Collagen I Inserts with PET Membrane
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
£338.12 - £470.80
Specifications
For Use With (Application) | Adhesion, growth and differentiation of neuronal and transfected cell lines |
---|---|
Storage Requirements | 2° to 8°C |
Product Code | Brand | Pore Size | Type | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Type | Quantity | Price | Quantity & Availability | |||
10133861
|
Corning™
354598 |
0.3 μm | Cell Culture Insert | 1 | N/A | ||||
10488262
|
Corning™
354482 |
1 μm | Well Plate | 2 | N/A | ||||
10366151
|
Corning™
354442 |
0.45 μm | Well Plate | 4 | N/A | ||||
10295091
|
Corning™
354444 |
0.4 μm | Well Plate | 2 |
£455.65
Pack of 24 |
In Stock
Log in to see stock. |
|||
10122851
|
Corning™
354540 |
3 μm | Well Plate | 4 | N/A | ||||
10526001
|
Corning™
354541 |
3 μm | Well Plate | 2 |
£440.14
Pack of 24 |
Estimated Shipment: 13-05-2025
Log in to see stock. |
|||
Description
- Each lot is tested for the ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
- Tested and found negative for bacteria and fungi
- Cellware is also tested for ability to promote firm attachment of rat cerebellar granule cells
- Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
- Matrix components are applied as an even, optically clear coating
- All lots tested and found negative for bacteria and fungi; cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
For research use only.
Order Info
Bulk quantities are also available; contact your Fisher Scientific Customer Service Representative.
Specifications
Adhesion, growth and differentiation of neuronal and transfected cell lines | |
2° to 8°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title