Learn More
Corning™ BioCoat™ Collagen I Multiwell Plates
Treated with BioCoat™ for enhanced cell attachment and growth of a variety of primary cells and ransformed cell lines in serum-free or reduced serum cultures. Corning™ BioCoat™ Collagen I Multiwell Plates closely mimmics in vivo environments for cell culture applications. Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.
Brand: Corning™ 356505
Code : 66
Additional Details : Weight : 4.37000kg
Description
Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.
Applications include:
- Promotion of cell attachment and spreading
- Rapid expansion of cell populations
- Serum-free or reduced serum culture
- Cell adhesion assays
- Improving survival of primary cells in culture
Source
Quality Control
- Tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
- Tested and found negative for bacteria and fungi
- Collagen I purity >90% by SDS-PAGE
Storage
Specifications
6 x 8 | |
Clear | |
Polystyrene | |
Clear | |
Polystyrene | |
Non-sterile | |
Multi-well Cell Culture Plate | |
Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture | |
50/Cs. | |
50/Cs. | |
4°C to 30°C |
ANSI, SBS | |
Yes | |
Flat | |
0.75 cm2 | |
Rectangular | |
Cell Culture | |
48 | |
Lid | |
5/Pk. | |
Rat Tail |