Learn More
Corning™ BioCoat™ 96-Well, Collagen Type I-Treated, Flat-Bottom Microplate
Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:With Lid
Brand: Corning™ 4582
Code : 67
Additional Details : Weight : 1.19000kg
Description
High optical quality, glass bottom, black microplates are ideal for performing high content cell-based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput, and is ideal for cell growth.
- High optical quality and scratch resistant glass
- Glass bottom thickness of 200 μm is well suited for imaging microscopy.
- Bottom flatness <50 μm to ensure planarity for imaging devices
- Low background fluorescence and minimal cross-talk provides the highest possible optical quality for cell-based assays.
- Half area 96-well microplate reduces reagent consumption.
Specifications
ANSI, SBS | |
With Lid | |
Black, Clear Bottom | |
96 | |
Collagen Type I | |
Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture | |
1/Pk. | |
Rat Tail |
Half-Area | |
8 x 12 | |
Polystyrene | |
Non-sterile | |
Round, Flat Bottom (F-well) | |
10/Cs. | |
10/Cs. | |
Clear |