missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ Collagen I, Rat Tail Culture Dish
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
£87.87 - £426.69
Specifications
Closure Color | Clear |
---|---|
Closure Included | Closure Included |
Closure Material | Polystyrene |
Bottom Shape | Round |
Color | Clear |
Product Code | Brand | Diameter (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Quantity | Price | Quantity & Availability | ||||
10555421
|
Corning™
356401 |
60 mm | 100/cs. |
£426.69
Pack of 100 |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
10772531
|
Corning™
356456 |
35 mm | 100/cs. |
£426.69
Pack of 100 |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
10155670
|
Corning™
354456 |
35 mm | 20/cs. |
£106.65
Pack of 20 |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
10543051
|
Corning™
354401 |
60 mm | 20/cs. |
£106.65
Pack of 20 |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
10572701
|
Corning™
354450 |
100 mm | 10/cs. |
£87.87
Pack of 10 |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
10185760
|
Corning™
354551 |
150 mm | 5/cs. |
£128.00
Pack of 5 |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
10499012
|
Corning™
356450 |
100 mm | 40/cs. |
£301.19
Pack of 40 |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
Description
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
- All lots tested and found negative for bacteria and fungi
- Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
Clear | |
Polystyrene | |
Clear | |
Round | |
Cell Culture Dish |
Closure Included | |
Round | |
Polystyrene | |
Collagen Type I | |
Lid |