Learn More
Corning™ BioCoat™ Fibronectin HTS Multiwell Angiogenesis
Provides a standard format to increase the quality and reproducibility of in vitro endothelial cell migration assays
£454.75 - £3705.00
Specifications
Pore Size | 3 μm |
---|---|
Type | Multiwell Insert Plate |
For Use With (Application) | High Throughput Screening |
Surface Coating | Fibronectin |
Includes: A Falcon 24- and 96-Multiwell™ insert plate containing a fluorescence blocking, microporous polyethylene terephthalate (PET) membrane evenly coated with human fibronectin, an untreated receiver plate, and a lid
Product Code | Brand | Culture Area | No. of Wells | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Culture Area | No. of Wells | Quantity | Price | Quantity & Availability | |||
10678892
|
Corning™
354144 |
0.3 cm2 | 24 | 5 |
£2038.44
Pack of 5 |
Estimated Shipment: 17-10-2024
Log in to see stock. |
|||
10067911
|
Corning™
354143 |
0.3 cm2 | 24 | 1 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10396602
|
Corning™
354148 |
0.08 cm2 | 96 | 5 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10778124
|
Corning™
354147 |
0.08 cm2 | 96 | 1 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
Specifications
3 μm | |
Multiwell Insert Plate | |
High Throughput Screening | |
Fibronectin |